Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rxrbb
Entrez Gene ID
30486
Gene Name
retinoid x receptor, beta b
Gene Aliases
RXR, rxrd, unp286
Chromosome Location
Chr.16: 166151 - 194337 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131238.1 NP_571313.1 6 - 7 849 105
XM_021466616.1 XP_021322291.1 6 - 7 826 105
XM_021466617.1 XP_021322292.1 6 - 7 828 105
GenBank mRNA BC162301.1 - 6 - 7 843 105
BC162302.1 - 6 - 7 843 105
U29941.1 - 6 - 7 849 105

View Full Product Details