Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rxrbb
Entrez Gene ID
30486
Gene Name
retinoid x receptor, beta b
Gene Aliases
RXR, rxrd, unp286
Chromosome Location
Chr.16: 166151 - 194337 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131238.1 NP_571313.1 9 - 10 1120 65
XM_021466616.1 XP_021322291.1 9 - 10 1097 65
XM_021466617.1 XP_021322292.1 9 - 10 1099 65
GenBank mRNA BC162301.1 - 9 - 10 1114 65
BC162302.1 - 9 - 10 1114 65
U29941.1 - 9 - 10 1120 65

View Full Product Details