Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dlx5a
Entrez Gene ID
30569
Gene Name
distal-less homeobox 5a
Gene Aliases
dlx4, zgc:101787
Chromosome Location
Chr.19: 41470216 - 41472228 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131306.2 NP_571381.2 1 - 2 534 89
GenBank mRNA BC083280.1 - 1 - 2 468 89
BC165702.1 - 1 - 2 396 89
EH449338.1 - 1 - 2 499 89
EH606041.1 - 2 - 3 766 89
U03876.1 - 1 - 2 459 89

View Full Product Details