Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
six4a
Entrez Gene ID
65233
Gene Name
SIX homeobox 4a
Gene Aliases
fk50f02, six4.2, six4b, wu:fk50f02
Chromosome Location
Chr.13: 31636728 - 31647006 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131718.2 NP_571793.2 2 - 3 1522 83
GenBank mRNA AB045623.1 - 2 - 3 1528 83
BC134897.1 - 2 - 3 1505 83

View Full Product Details