Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ngb
Entrez Gene ID
100000711
Gene Name
neuroglobin
Gene Aliases
-
Chromosome Location
Chr.17: 390191 - 396410 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131853.1 NP_571928.1 2 - 3 297 64
XM_021467082.1 XP_021322757.1 2 - 3 425 64
GenBank mRNA AJ315610.1 - 2 - 3 297 64
BC059416.1 - 2 - 3 289 64

View Full Product Details