Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dacha
Entrez Gene ID
192121
Gene Name
dachshund a
Gene Aliases
cb907
Chromosome Location
Chr.21: 34363972 - 34658298 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152955.2 NP_694487.2 3 - 4 882 98
XM_005161406.4 XP_005161463.1 3 - 4 909 98
XM_009295592.3 XP_009293867.1 3 - 4 909 98
XM_009295593.3 XP_009293868.1 3 - 4 909 98
XM_021468748.1 XP_021324423.1 3 - 4 909 98
GenBank mRNA AF427108.1 - 3 - 4 883 98
BC162471.1 - 3 - 4 795 98

View Full Product Details