Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dbr1
Entrez Gene ID
323746
Gene Name
debranching RNA lariats 1
Gene Aliases
wu:fc08f08, zgc:63930
Chromosome Location
Chr.9: 527315 - 535245 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199653.1 NP_955947.1 3 - 4 534 79
GenBank mRNA BC053153.1 - 3 - 4 534 79

View Full Product Details