Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
n4bp1
Entrez Gene ID
327327
Gene Name
nedd4 binding protein 1
Gene Aliases
wu:fi03b05, zgc:66437
Chromosome Location
Chr.18: 18406033 - 18422543 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199787.2 NP_956081.2 6 - 7 2469 77
GenBank mRNA BC059205.1 - 6 - 7 2469 77

View Full Product Details