Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rasl11b
Entrez Gene ID
393109
Gene Name
RAS-like, family 11, member B
Gene Aliases
fc21f04, wu:fc21f04, zgc:55750, zgc:77749
Chromosome Location
Chr.20: 23022985 - 23026223 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200140.1 NP_956434.1 2 - 3 357 63
GenBank mRNA BC044471.1 - 2 - 3 357 63
BC065446.1 - 2 - 3 328 63
DQ983377.1 - 2 - 3 341 63

View Full Product Details