Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dcp2
Entrez Gene ID
393121
Gene Name
decapping mRNA 2
Gene Aliases
zgc:55938, zgc:77040
Chromosome Location
Chr.10: 6473092 - 6494039 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200152.1 NP_956446.1 6 - 7 897 83
GenBank mRNA BC044547.1 - 6 - 7 897 83
BC066577.1 - 6 - 7 928 83

View Full Product Details