Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dcp2
Entrez Gene ID
393121
Gene Name
decapping mRNA 2
Gene Aliases
zgc:55938, zgc:77040
Chromosome Location
Chr.10: 6473092 - 6494039 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200152.1 NP_956446.1 7 - 8 999 66
GenBank mRNA BC044547.1 - 7 - 8 999 66

View Full Product Details