Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd2l
Entrez Gene ID
393192
Gene Name
programmed cell death 2-like
Gene Aliases
zgc:56288
Chromosome Location
Chr.25: 30442111 - 30454774 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200223.1 NP_956517.1 1 - 2 172 113
GenBank mRNA BC046049.1 - 1 - 2 172 113

View Full Product Details