Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdcd2l
Entrez Gene ID
393192
Gene Name
programmed cell death 2-like
Gene Aliases
zgc:56288
Chromosome Location
Chr.25: 30442111 - 30454774 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200223.1 NP_956517.1 4 - 5 853 87
XM_021470458.1 XP_021326133.1 4 - 5 1705 87
XM_021470459.1 XP_021326134.1 4 - 5 1692 87
XM_021470460.1 XP_021326135.1 4 - 5 1236 87
GenBank mRNA BC046049.1 - 4 - 5 853 87

View Full Product Details