Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnaseh2a
Entrez Gene ID
393195
Gene Name
ribonuclease H2, subunit A
Gene Aliases
zgc:56307
Chromosome Location
Chr.1: 51712872 - 51720632 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200226.2 NP_956520.2 5 - 6 625 98
GenBank mRNA AY391457.1 - 4 - 5 424 98
BC046061.1 - 5 - 6 451 98
BC071375.1 - 5 - 6 625 98

View Full Product Details