Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc25a32a
Entrez Gene ID
393226
Gene Name
solute carrier family 25 member 32a
Gene Aliases
fi40c12, mftc, wu:fi40c12, zgc:55610
Chromosome Location
Chr.19: 12937069 - 12965589 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200256.1 NP_956550.1 3 - 4 637 83
GenBank mRNA BC048057.1 - 3 - 4 637 83
BC164127.1 - 3 - 4 465 83

View Full Product Details