Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc35d1b
Entrez Gene ID
393447
Gene Name
solute carrier family 35 member D1b
Gene Aliases
zgc:63651
Chromosome Location
Chr.2: 5793875 - 5810326 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200475.2 NP_956769.2 6 - 7 603 106
GenBank mRNA BC055186.1 - 6 - 7 603 106

View Full Product Details