Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zgpat
Entrez Gene ID
393457
Gene Name
zinc finger, CCCH-type with G patch domain
Gene Aliases
zgc:63730
Chromosome Location
Chr.11: 12860499 - 12872409 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200485.1 NP_956779.1 2 - 3 659 73
GenBank mRNA BC055224.1 - 2 - 3 659 73

View Full Product Details