Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
efna1a
Entrez Gene ID
393569
Gene Name
ephrin-A1a
Gene Aliases
zgc:63493
Chromosome Location
Chr.19: 8481802 - 8520883 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200597.2 NP_956891.1 4 - 5 731 61
GenBank mRNA BC056770.1 - 4 - 5 731 61

View Full Product Details