Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2e3
Entrez Gene ID
321689
Gene Name
ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast)
Gene Aliases
UbcD2, wu:fb26h04, zgc:55923, zgc:76906
Chromosome Location
Chr.9: 43797902 - 43880341 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200921.3 NP_957215.2 5 - 6 887 88
GenBank mRNA BC042331.1 - 5 - 6 649 88
BC067146.1 - 5 - 6 639 88

View Full Product Details