Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dynlt3
Entrez Gene ID
402901
Gene Name
dynein, light chain, Tctex-type 3
Gene Aliases
tcte1l, zgc:101678
Chromosome Location
Chr.11: 29790626 - 29794308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205608.1 NP_991171.1 4 - 5 351 87
GenBank mRNA AY398423.1 - 4 - 5 351 87

View Full Product Details