Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc35e1
Entrez Gene ID
406347
Gene Name
solute carrier family 35 member E1
Gene Aliases
fc05g11, fc55b08, wu:fc05g11, wu:fc55b08, zgc:55838
Chromosome Location
Chr.11: 5499614 - 5512719 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213074.1 NP_998239.1 3 - 4 591 85
GenBank mRNA BC046896.1 - 3 - 4 591 85

View Full Product Details