Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
galcb
Entrez Gene ID
406385
Gene Name
galactosylceramidase b
Gene Aliases
galc, wu:fi04e07, zgc:56444
Chromosome Location
Chr.17: 18889637 - 18898082 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213111.1 NP_998276.1 9 - 10 1038 60
GenBank mRNA BC049316.1 - 9 - 10 1038 60

View Full Product Details