Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ola1
Entrez Gene ID
326864
Gene Name
Obg-like ATPase 1
Gene Aliases
fe36h01, wu:fc66b09, wu:fe36h01, wu:fk82a02, zgc:55768, zgc:85691
Chromosome Location
Chr.6: 10741266 - 10753507 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281986.1 NP_001268915.1 3 - 4 374 140
NM_214700.2 NP_999865.1 4 - 5 518 140
XM_021476786.1 XP_021332461.1 4 - 5 484 140
GenBank mRNA BC051155.1 - 4 - 5 466 140
BC067579.1 - 4 - 5 451 140
CT720868.1 - 3 - 4 314 140
EH542806.1 - 4 - 5 506 140
EH578941.1 - 4 - 5 534 140

View Full Product Details