Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ola1
Entrez Gene ID
326864
Gene Name
Obg-like ATPase 1
Gene Aliases
fe36h01, wu:fc66b09, wu:fe36h01, wu:fk82a02, zgc:55768, zgc:85691
Chromosome Location
Chr.6: 10741266 - 10753507 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281986.1 NP_001268915.1 7 - 8 866 113
NM_214700.2 NP_999865.1 8 - 9 1010 113
XM_021476786.1 XP_021332461.1 8 - 9 976 113
GenBank mRNA BC051155.1 - 8 - 9 958 113
BC067579.1 - 8 - 9 943 113
EH611446.1 - 3 - 4 531 113

View Full Product Details