Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmgn3
Entrez Gene ID
492345
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
fa04b03, trip7, wu:fa04b03, zgc:92355
Chromosome Location
Chr.23: 31358085 - 31372807 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007311.2 NP_001007312.1 6 - 6 505 119
NM_001243173.1 NP_001230102.1 7 - 7 605 119
NM_001243174.1 NP_001230103.1 7 - 7 602 119
NM_001243176.1 NP_001230105.1 6 - 6 502 119
GenBank mRNA BC085665.1 - 6 - 6 118
BC152615.1 - 6 - 6 446 119
EH439229.1 - 6 - 6 425 119
EH475164.1 - 1 - 1 625 119
EH590201.1 - 6 - 6 507 119

View Full Product Details