Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smcr8b
Entrez Gene ID
407723
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8b
Gene Aliases
smcr8
Chromosome Location
Chr.1: 1 - 4529 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214802.1 NP_999967.1 1 - 1 853 143
GenBank mRNA AY576998.1 - 1 - 1 853 143
BC162340.1 - 1 - 1 607 143

View Full Product Details