Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pkz
Entrez Gene ID
503703
Gene Name
protein kinase containing Z-DNA binding domains
Gene Aliases
zgc:136657
Chromosome Location
Chr.13: 50189890 - 50200119 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040376.2 NP_001035466.1 11 - 11 2448 127
GenBank mRNA BC115215.1 - 11 - 11 2412 127
BC131857.1 - 11 - 11 2448 127

View Full Product Details