Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
st3gal7
Entrez Gene ID
554166
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 7
Gene Aliases
siat5-related, st3GalV.2, st3gal5l, wu:fb60h07, zgc:153853
Chromosome Location
Chr.13: 40716318 - 40726881 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004016.2 NP_001004016.2 7 - 7 1188 130
XM_005157060.4 XP_005157117.1 7 - 7 1201 130
GenBank mRNA AJ783742.1 - 6 - 6 1050 130
BC124750.1 - 7 - 7 1161 130

View Full Product Details