The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017567.2 | NP_001017567.1 | 5 - 5 | 1054 | 125 |
GenBank mRNA | BC092748.1 | - | 5 - 5 | 1044 | 125 |
EU368947.1 | - | 5 - 5 | 1035 | 125 |