Assay Design
The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Gene Transcripts
- Gene Symbol
- mpz
- Entrez Gene ID
-
114417
- Gene Name
- myelin protein zero
- Gene Aliases
-
p0, sc:d0186, wu:fc04b11, wu:fi30g06, zgc:103775
- Chromosome Location
-
Chr.2: 176690 - 198655 on Build GRCz11
- Gene Symbol
- grb10a
- Entrez Gene ID
-
446167
- Gene Name
- growth factor receptor-bound protein 10a
- Gene Aliases
-
grb10, zgc:91987
- Chromosome Location
-
Chr.19: 23136170 - 23249849 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC080269.1 |
- |
- |
3152 |
76 |
View Full Product Details