Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fign
Entrez Gene ID
553599
Gene Name
fidgetin
Gene Aliases
zgc:110229
Chromosome Location
Chr.9: 50599362 - 50753942 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020575.1 NP_001018411.1 2 - 2 2950 102
XM_021478666.1 XP_021334341.1 2 - 2 2973 102
GenBank mRNA BC095207.1 - 2 - 2 2950 102

View Full Product Details