Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pcdh2g1
Entrez Gene ID
554047
Gene Name
protocadherin 2 gamma 1
Gene Aliases
-
Chromosome Location
Chr.14: 1889679 - 2097719 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024174.1 NP_001019345.1 1 - 1 457 142
GenBank mRNA AY583034.1 - 1 - 1 457 142
BC162458.1 - 1 - 1 457 142

View Full Product Details