Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pcdh2g5
Entrez Gene ID
554053
Gene Name
protocadherin 2 gamma 5
Gene Aliases
-
Chromosome Location
Chr.14: 1889679 - 2077281 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024178.1 NP_001019349.1 1 - 1 51 100
GenBank mRNA AY583053.1 - 1 - 1 51 100

View Full Product Details