Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taar14i
Entrez Gene ID
100034374
Gene Name
trace amine associated receptor 14i
Gene Aliases
si:dkey-246e3.4, taar68
Chromosome Location
Chr.20: 46255056 - 46256016 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083037.1 NP_001076506.1 1 - 1 453 81

View Full Product Details