Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
txlna
Entrez Gene ID
324180
Gene Name
taxilin alpha
Gene Aliases
fc21b12, im:7039434, wu:fc21b12, zgc:162882
Chromosome Location
Chr.19: 29836458 - 29853410 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099230.1 NP_001092700.1 11 - 11 2753 86
XM_017352186.2 XP_017207675.1 10 - 10 2699 86
GenBank mRNA BC139888.1 - 11 - 11 2753 86

View Full Product Details