Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
foxg1a
Entrez Gene ID
30274
Gene Name
forkhead box G1a
Gene Aliases
BF-1, bf1, foxg1, zgc:109850
Chromosome Location
Chr.17: 29104157 - 29119470 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131067.1 NP_571142.1 1 - 1 2153 135
GenBank mRNA AF067204.1 - 1 - 1 2153 135

View Full Product Details