Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
etv6
Entrez Gene ID
114444
Gene Name
ETS variant transcription factor 6
Gene Aliases
id:ibd2308, tel
Chromosome Location
Chr.Unknown: 5152 - 5447 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131832.2 NP_571907.2 - 1753 110
GenBank mRNA AF339838.1 - - 1539 110
AY693994.1 - - 1549 110
BC045451.1 - - 1753 110
BC065661.1 - - 1750 110

Gene Symbol
rhobtb2b
Entrez Gene ID
565844
Gene Name
Rho related BTB domain containing 2b
Gene Aliases
-
Chromosome Location
Chr.10: 42689941 - 42714941 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017358113.2 XP_017213602.2 - 3463 110

View Full Product Details