Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcdh1g22
Entrez Gene ID
108191476
Gene Name
protocadherin 1 gamma 22
Gene Aliases
fc15h06, fj64d07, wu:fc15h06, wu:fj64d07, zgc:73319
Chromosome Location
Chr.10: 21776760 - 21779594 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY576949.1 - 1 - 1 1557 150
BC066125.1 - 1 - 1 1646 150

View Full Product Details