Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
timp2b
Entrez Gene ID
406650
Gene Name
TIMP metallopeptidase inhibitor 2b
Gene Aliases
timp2l, zgc:65967, zgc:85799
Chromosome Location
Chr.3: 57632965 - 57666518 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213296.1 NP_998461.1 5 - 5 850 91
GenBank mRNA BC056534.1 - 5 - 5 807 91
BC068388.1 - 5 - 5 850 91

View Full Product Details