Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prdx2
Entrez Gene ID
791455
Gene Name
peroxiredoxin 2
Gene Aliases
cb58, prdx1, wu:fq19b09, zgc:92891
Chromosome Location
Chr.1: 51721851 - 51729866 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002468.2 NP_001002468.1 4 - 5 489 150
GenBank mRNA BC076347.1 - 4 - 5 445 150
EH486177.1 - 4 - 5 499 150
EH575743.1 - 2 - 3 548 150

View Full Product Details