Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
suz12a
Entrez Gene ID
794171
Gene Name
SUZ12 polycomb repressive complex 2 subunit a
Gene Aliases
suz12, suz12l, zgc:100863
Chromosome Location
Chr.3: 35849672 - 35865795 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003529.2 NP_001003529.1 9 - 10 1147 73
XM_021471664.1 XP_021327339.1 9 - 10 2162 73
GenBank mRNA BC078293.1 - 9 - 10 1150 73
BC152625.1 - 9 - 10 1036 73

View Full Product Details