Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trappc3
Entrez Gene ID
445207
Gene Name
trafficking protein particle complex 3
Gene Aliases
zgc:101005
Chromosome Location
Chr.19: 35722902 - 35733383 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003601.1 NP_001003601.1 2 - 3 251 105
GenBank mRNA BC078259.1 - 2 - 3 251 105

View Full Product Details