Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plk1
Entrez Gene ID
280649
Gene Name
polo-like kinase 1 (Drosophila)
Gene Aliases
cb525, cb636, ik:tdsubc_2d1, plk, wu:fb37g11, wu:fb76g03, xx:tdsubc_2d1
Chromosome Location
Chr.1: 9153187 - 9161695 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003890.3 NP_001003890.3 10 - 11 1867 74
GenBank mRNA AY648752.1 - 10 - 11 1826 74
BC098545.1 - 10 - 11 1867 74
HM210100.1 - 10 - 11 1583 74

View Full Product Details