Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arrdc1a
Entrez Gene ID
100006005
Gene Name
arrestin domain containing 1a
Gene Aliases
arrdc1, sb:cb786, si:rp71-1o1.5, wu:fc25g09, wu:fi04h05, zgc:92248
Chromosome Location
Chr.5: 29516843 - 29531948 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004561.1 NP_001004561.1 5 - 6 701 78
XM_005171783.4 XP_005171840.1 5 - 6 686 78
GenBank mRNA BC081631.1 - 5 - 6 655 78
BC096962.1 - 5 - 6 680 78
BC152620.1 - 5 - 6 670 78
BC165815.1 - 5 - 6 649 78

View Full Product Details