Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nucks1a
Entrez Gene ID
447862
Gene Name
nuclear casein kinase and cyclin-dependent kinase substrate 1a
Gene Aliases
nucks1, wu:fb16a10, wu:fb16h08, wu:fb36b01, wu:fb82a01, wu:fe37f10, wu:gcd26e12, zgc:86878
Chromosome Location
Chr.11: 38540138 - 38553770 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004601.2 NP_001004601.1 3 - 4 406 100
GenBank mRNA AI588726.1 - 3 - 4 406 100
AI626711.1 - 3 - 4 363 100
BC081557.1 - 3 - 4 381 100
BC154185.1 - 3 - 4 392 100
CT705076.2 - 2 - 3 125 100

View Full Product Details