Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nucks1a
Entrez Gene ID
447862
Gene Name
nuclear casein kinase and cyclin-dependent kinase substrate 1a
Gene Aliases
nucks1, wu:fb16a10, wu:fb16h08, wu:fb36b01, wu:fb82a01, wu:fe37f10, wu:gcd26e12, zgc:86878
Chromosome Location
Chr.11: 38540138 - 38553770 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004601.2 NP_001004601.1 5 - 6 600 94
GenBank mRNA BC081557.1 - 5 - 6 575 94
BC154185.1 - 5 - 6 586 94
CT705076.2 - 4 - 5 319 94

View Full Product Details