Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpd1l
Entrez Gene ID
449663
Gene Name
glycerol-3-phosphate dehydrogenase 1 like
Gene Aliases
wu:fi13g03, wu:fi45b08, zgc:92580
Chromosome Location
Chr.16: 39248644 - 39267185 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005934.2 NP_001005934.1 7 - 8 1064 80
GenBank mRNA BC083522.1 - 7 - 8 1044 80

View Full Product Details