Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tspan13b
Entrez Gene ID
449797
Gene Name
tetraspanin 13b
Gene Aliases
tspan13, zgc:101833
Chromosome Location
Chr.19: 30387996 - 30399203 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005970.1 NP_001005970.1 3 - 4 497 68
GenBank mRNA BC083295.1 - 3 - 4 497 68
BC165008.1 - 3 - 4 341 68

View Full Product Details