Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adprhl1
Entrez Gene ID
450012
Gene Name
ADP-ribosylhydrolase like 1
Gene Aliases
zgc:103446
Chromosome Location
Chr.1: 197236 - 206299 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006033.2 NP_001006033.1 4 - 5 687 71
XM_005167996.4 XP_005168053.1 4 - 5 684 71
XM_021473006.1 XP_021328681.1 4 - 5 778 71
GenBank mRNA BC083385.1 - 4 - 5 687 71

View Full Product Details