Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptpn4a
Entrez Gene ID
368448
Gene Name
protein tyrosine phosphatase non-receptor type 4a
Gene Aliases
ptpn4, si:busm1-37o5.1, si:dz37o5.1
Chromosome Location
Chr.9: 220637 - 276592 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007200.1 NP_001007201.1 12 - 13 1086 71

View Full Product Details